Phenyl trifluoromethyl sulphoxide ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: ((Trifluoromethyl)sulphinyl)benzene, [(Trifluoromethyl)sulphinyl]benzene

P2460-1G P2460-5G
TCIAP2460-1GEA 1325 SEK
TCIAP2460-1G TCIAP2460-5G
Phenyl trifluoromethyl sulphoxide ≥98.0%
Phenyl trifluoromethyl sulphoxide
Formel: C₇H₅F₃OS
MW: 194,17 g/mol
Kokpunkt: 82°C
MDL Number: MFCD30475661
CAS nummer: 703-18-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=916a0b1c0f538cf9893be640d6c08405b9aa5562&vid=916a0b1c0f538cf9893be640d6c08405b9aa5562&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR