1-Propoxy-4-[[4-(trans-4-propylcyclohexyl)phenyl]ethynyl]benzene ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 1-[(4-Propoxyphenyl)ethynyl]-4-(trans-4-propylcyclohexyl)benzene

P2440-1G P2440-5G
TCIAP2440-1GEA 750 SEK
TCIAP2440-1G TCIAP2440-5G
1-Propoxy-4-[[4-(trans-4-propylcyclohexyl)phenyl]ethynyl]benzene ≥98.0%
1-Propoxy-4-[[4-(trans-4-propylcyclohexyl)phenyl]ethynyl]benzene
Formel: C₂₆H₃₂O
MW: 360,54 g/mol
MDL Number: MFCD11053434
CAS nummer: 116903-49-2

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Mesomorphic range 96 to 233 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR