L-Prolinanilide ≥98.0% (by GC, titration analysis)

Leverantör: TCI
Varning

P2430-1G P2430-5G
TCIAP2430-1GEA 855 SEK
TCIAP2430-1G TCIAP2430-5G
L-Prolinanilide ≥98.0% (by GC, titration analysis)
L-Prolinanilide
Formel: C₁₁H₁₄N₂O
MDL Number: MFCD08276245
CAS nummer: 64030-43-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=823c8b7119f98a9b5c91a7832ec89d506866dc4e&vid=823c8b7119f98a9b5c91a7832ec89d506866dc4e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 76.0 to 80.0 deg-C
Specific rotation [a]20/D -65.0 to -70.0 deg (C=1, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR