3-Phenyl-9H-carbazole ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
P2401-1G
P2401-5G
TCIAP2401-1GEA
350
SEK
TCIAP2401-1G
TCIAP2401-5G
3-Phenyl-9H-carbazole ≥98.0%
3-Phenyl-9H-carbazole
Formel:
C₁₈H₁₃N MW: 243,31 g/mol |
MDL Number:
MFCD22052094 CAS nummer: 103012-26-6 |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 220.0 to 224.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...