1-(4-Propoxyphenyl)ethanone ≥98.0%
Leverantör: TCI EUROPE
Synonyms: 4'-Propoxyacetophenone
P2377-1G P2377-5G
TCIAP2377-1GEA 830 SEK
TCIAP2377-1G TCIAP2377-5G
1-(4-Propoxyphenyl)ethanone ≥98.0%
1-(4-Propoxyphenyl)ethanone
Formel: C₁₁H₁₄O₂
MW: 178,23 g/mol
Kokpunkt: 111°C
Storage Temperature: Kylskåp
MDL Number: MFCD01922144
CAS nummer: 5736-86-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d15f7bb6b596f30291e4bfd08c73cf0e4d0ca612&vid=d15f7bb6b596f30291e4bfd08c73cf0e4d0ca612&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %