4-[2-(trans-4-Propylcyclohexyl)ethyl]cyclohexanone ≥98.0%

Leverantör: TCI EUROPE

Synonyms: trans-4-[2-(4-Propylcyclohexyl)ethyl]cyclohexanone

P2372-1G P2372-5G
TCIAP2372-1GEA 630 SEK
TCIAP2372-1G TCIAP2372-5G
4-[2-(trans-4-Propylcyclohexyl)ethyl]cyclohexanone ≥98.0%
4-[2-(trans-4-Propylcyclohexyl)ethyl]cyclohexanone
Formel: C₁₇H₃₀O
MW: 250,43 g/mol
MDL Number: MFCD12911604
CAS nummer: 117923-32-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12ace664fdcbff963c685d9d05090ec6b2276ffc&vid=12ace664fdcbff963c685d9d05090ec6b2276ffc&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR