9-Phenyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 9-Phenylcarbazole-2-boronic acid Pinacol ester, 9-Phenyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole, 4,4,5,5-Tetramethyl-2-(9-phenylcarbazol-2-yl)-1,3,2-dioxaborolane

P2364-200MG P2364-1G
TCIAP2364-200MGEA 485 SEK
TCIAP2364-200MG TCIAP2364-1G
9-Phenyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole ≥98.0%
9-Phenyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
Formel: C₂₄H₂₄BNO₂
MW: 369,27 g/mol
MDL Number: MFCD28129441
CAS nummer: 1246669-45-3

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 147.0 to 151.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR