3-Phosphonopropionic acid ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 3-Phosphonopropanoic acid

P2348-1G P2348-5G
TCIAP2348-1GEA 270 SEK
TCIAP2348-1G TCIAP2348-5G
3-Phosphonopropionic acid ≥98.0%
3-Phosphonopropionic acid
Formel: C₃H₇O₅P
MW: 154,06 g/mol
MDL Number: MFCD00002770
CAS nummer: 5962-42-5
EINECS: 227-738-8
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=825c67a8c997c7833306f4513fccb5330092bf9a&vid=825c67a8c997c7833306f4513fccb5330092bf9a&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystaline
Purity (Neutralization titration) min. 98.0 %
Melting point 166.0 to 170.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR