(Phenylhydrazono)malononitrile ≥96.0%

Leverantör: TCI EUROPE
Fara

P2346-5G P2346-25G
TCIAP2346-5GEA 670 SEK
TCIAP2346-5G TCIAP2346-25G
(Phenylhydrazono)malononitrile ≥96.0%
(Phenylhydrazono)malononitrile
Formel: C₉H₆N₄
MDL Number: MFCD00195983
CAS nummer: 306-18-3
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45c19440c46f8c0723a695b919adff694a08b5e8&vid=9fa58543da1dbe800c9b5f7592a40871f0fb2ad0&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 96.0 %
Melting point 131.0 to 135.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR