N-(Propargyloxy)phthalimide ≥98.0%

Leverantör: TCI
Varning

P2342-1G P2342-5G
TCIAP2342-1GEA 510 SEK
TCIAP2342-1G TCIAP2342-5G
N-(Propargyloxy)phthalimide ≥98.0%
N-(Propargyloxy)phthalimide
Formel: C₁₁H₇NO₃
MDL Number: MFCD00005890
CAS nummer: 4616-63-1
EINECS: 225-024-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6879a7caeb1a385347419294915c275730ecad1&vid=c6879a7caeb1a385347419294915c275730ecad1&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 138.0 to 142.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR