Propargyl sulphide ≥98.0%
Leverantör: TCI
Varning
Synonyms: Dipropargyl sulphide
P2336-1G P2336-5G
TCIAP2336-1GEA 1600 SEK
TCIAP2336-1G TCIAP2336-5G
Propargyl sulphide ≥98.0%
Propargyl sulphide
Formel: C₆H₆S
MW: 110.17 g/mol
Kokpunkt: 59°C
Storage Temperature: Kylskåp
MDL Number: MFCD00048109
CAS nummer: 13702-09-5
UN: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7b04f9e5e6a50504c6e22c3c7d900b5817ef5ec&vid=b06c461835133c5ceaea28162887f0b7f542b6d9&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %