Propargyl ether ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: Dipropargylether
P2335-1G P2335-5G
TCIAP2335-1GEA 830 SEK
TCIAP2335-1G TCIAP2335-5G
Propargyl ether ≥98.0%
Propargyl ether
Formel: C₆H₆O
MW: 94,11 g/mol
Kokpunkt: 117°C
Flampunkt: 23°C
Storage Temperature: Kylskåp
MDL Number: MFCD00048108
CAS nummer: 6921-27-3
UN: 3271
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a966c29ab3d96689d6779d862695cdfb3ed9365&vid=3a966c29ab3d96689d6779d862695cdfb3ed9365&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %