trans,trans-4-Pentyl-4'-vinylbicyclohexyl ≥98.0%
Leverantör: TCI EUROPE
Synonyms: trans,trans-4-Amyl-4'-vinylbicyclohexyl
P2318-1G P2318-5G
TCIAP2318-1GEA 365 SEK
TCIAP2318-1G TCIAP2318-5G
trans,trans-4-Pentyl-4'-vinylbicyclohexyl ≥98.0%
trans,trans-4-Pentyl-4'-vinylbicyclohexyl
Formel: C₁₉H₃₄
MW: 262,48 g/mol
MDL Number: MFCD23135326
CAS nummer: 129738-34-7
Order Now
Specifikation testresultat
Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Melting point 47.0 to 51.0 deg-C