Pipamperone ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
1'-[4-(4-Fluorophenyl)-4-oxobutyl]-[1,4'-bipiperidine]-4'-carboxamide, Floropipamide
P2315-25MG
P2315-100MG
TCIAP2315-25MGEA
845
SEK
TCIAP2315-25MG
TCIAP2315-100MG
Pipamperone ≥98.0%
Pipamperone
Formel:
C₂₁H₃₀FN₃O₂ MW: 375,49 g/mol Smältpunkt: 126°C |
MDL Number:
MFCD00242979 CAS nummer: 1893-33-0 UN: 2811 ADR: 6.1,III Merck Index: 14,07455 |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 10.60 to 11.70 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...