Paclobutrazol ≥98.0%

Leverantör: TCI
Fara

P2299-1G P2299-5G
TCIAP2299-1GEA 255 SEK
TCIAP2299-1G TCIAP2299-5G
Paclobutrazol ≥98.0%
Paclobutrazol
Formel: C₁₅H₂₀ClN₃O
MDL Number: MFCD01678673
CAS nummer: 76738-62-0
UN: 1325
ADR: 4.1,II
Merck Index: 14,06984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=961a3d0d20117767cff1bef47e603d55b7ab7f5c&vid=961a3d0d20117767cff1bef47e603d55b7ab7f5c&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 161.0 to 166.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR