Pentafluoro(phenoxy)cyclotriphosphazene ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
2,4,4,6,6-Pentafluoro-2-phenoxy-1,3,5,2,4,6-triazatriphosphorine
P2286-1G
P2286-5G
TCIAP2286-1GEA
1250
SEK
TCIAP2286-1G
TCIAP2286-5G
Pentafluoro(phenoxy)cyclotriphosphazene ≥98.0%
Pentafluoro(phenoxy)cyclotriphosphazene
Formel:
C₆H₅F₅N₃OP₃ MW: 323,04 g/mol Kokpunkt: 46°C Storage Temperature: Kylskåp |
CAS nummer:
33027-68-8 UN: 1760 ADR: 8,III |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...