1-Phenylisatin ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: N-Phenylindoline-2,3-dione, 1-Phenylindoline-2,3-dione, N-Phenylisatin, N-Phenyl-1H-indole-2,3-dione, 1-Phenyl-2,3-dihydroindole-2,3-dione, N-Phenyl-2,3-dihydroindole-2,3-dione, 1-Phenyl-1H-indole-2,3-dione

P2279-5G P2279-25G
TCIAP2279-5GEA 1550 SEK
TCIAP2279-5G TCIAP2279-25G
1-Phenylisatin ≥98.0%
1-Phenylisatin
Formel: C₁₄H₉NO₂
MW: 223,23 g/mol
MDL Number: MFCD00082681
CAS nummer: 723-89-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a00675567d8e5991ebd28214b396a2cb8e2a08a&vid=6a00675567d8e5991ebd28214b396a2cb8e2a08a&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 139.0 to 143.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR