Phenyl acrylate ≥98.0% stabilised
Leverantör: TCI EUROPE
|
Varning
|
Stabilization:
Stabilised med BHT/Ionol
P2268-5G
P2268-25G
TCIAP2268-5GEA
630
SEK
TCIAP2268-5G
TCIAP2268-25G
Phenyl acrylate ≥98.0% stabilised
Phenyl acrylate
Formel:
C₉H₈O₂ Kokpunkt: 98°C |
MDL Number:
MFCD00048145 CAS nummer: 937-41-7 EINECS: 213-329-1 |
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...