3-Phenoxypropan-1-ol ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 3-Phenoxy-1-propanol

P2254-5G P2254-25G
TCIAP2254-5GEA 495 SEK
TCIAP2254-5G TCIAP2254-25G
3-Phenoxypropan-1-ol ≥98.0%
3-Phenoxypropan-1-ol
Formel: C₉H₁₂O₂
MW: 152,193 g/mol
Kokpunkt: 153°C
MDL Number: MFCD00039549
CAS nummer: 6180-61-6
EINECS: 228-228-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1c2e64e3791826c06db7c2d23f53f4c8c469b7d&vid=e1c2e64e3791826c06db7c2d23f53f4c8c469b7d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR