1-Phenylcyclobutanecarbonitrile ≥98.0%

Leverantör: TCI EUROPE
Varning

P2252-5G P2252-25G
TCIAP2252-5GEA 775 SEK
TCIAP2252-5G TCIAP2252-25G
1-Phenylcyclobutanecarbonitrile ≥98.0%
1-Phenylcyclobutanecarbonitrile
Formel: C₁₁H₁₁N
MW: 157,216 g/mol
Kokpunkt: 85°C
MDL Number: MFCD00019258
CAS nummer: 14377-68-5
EINECS: 238-351-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d06f07e60354fff4f0b5cce00932f10483989dd&vid=2d06f07e60354fff4f0b5cce00932f10483989dd&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR