1-(2-Propynyloxy)naphthalene ≥95.0%
Leverantör: TCI EUROPE
Synonyms: 1-(Propargyloxy)naphthalene
P2227-1G P2227-5G
TCIAP2227-1GEA 1050 SEK
TCIAP2227-1G TCIAP2227-5G
1-(2-Propynyloxy)naphthalene ≥95.0%
1-(2-Propynyloxy)naphthalene
Formel: C₁₃H₁₀O
MW: 182,22 g/mol
Kokpunkt: 75°C
Storage Temperature: Kylskåp
MDL Number: MFCD03787730
CAS nummer: 18542-45-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=895d15e7d0ed08073650f57aca70a0d2a76e7e86&vid=895d15e7d0ed08073650f57aca70a0d2a76e7e86&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %