1-[(2-Propynyloxy)methyl]pyrene ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 1-((Prop-2-yn-1-yloxy)methyl)pyrene, 1-[(Propargyloxy)methyl]pyrene

P2226-200MG P2226-1G
TCIAP2226-200MGEA 2025 SEK
TCIAP2226-200MG TCIAP2226-1G
1-[(2-Propynyloxy)methyl]pyrene ≥98.0%
1-[(2-Propynyloxy)methyl]pyrene
Formel: C₂₀H₁₄O
MW: 270,33 g/mol
MDL Number: MFCD28384153
CAS nummer: 1115084-83-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4a184360d8fa70e2d3f985abbe4d528ac58d266&vid=e4a184360d8fa70e2d3f985abbe4d528ac58d266&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 83.0 to 87.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR