Phenyl propargyl ether ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: (Prop-2-ynyloxy)benzene, (Prop-2-yn-1-yloxy)benzene
P2222-5G P2222-25G
TCIAP2222-5GEA 775 SEK
TCIAP2222-5G TCIAP2222-25G
Phenyl propargyl ether ≥98.0%
Phenyl propargyl ether
Formel: C₉H₈O
MW: 132,16 g/mol
Kokpunkt: 83°C
Flampunkt: 76°C
Storage Temperature: Kylskåp
MDL Number: MFCD00048107
CAS nummer: 13610-02-1
EINECS: 237-095-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=128f17a99f8961faac10bce30d4939764e4015e9&vid=128f17a99f8961faac10bce30d4939764e4015e9&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %