Potassium-5-methyl-1,3,4-oxadiazole-2-carboxylate ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE

Synonyms: 5-Methyl-1,3,4-oxadiazole-2-carboxylic acid potassium salt, Potassium 5-methyl-1,3,4-oxadiazole-2-carboxylate

P2220-200MG P2220-1G
TCIAP2220-200MGEA 1050 SEK
TCIAP2220-200MG TCIAP2220-1G
Potassium-5-methyl-1,3,4-oxadiazole-2-carboxylate ≥98.0% (by HPLC, titration analysis)
Potassium-5-methyl-1,3,4-oxadiazole-2-carboxylate
Formel: C4H3KN2O3
MW: 166,1800 g/mol
MDL Number: MFCD08060085
CAS nummer: 888504-28-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cdc80efca6d9392f0a6056456559ff381c102cf9&vid=cdc80efca6d9392f0a6056456559ff381c102cf9&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR