(1S)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline 98,0 GC, T
Leverantör: TCI
Synonyms:
(S)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline
P2216-1G
P2216-5G
TCIAP2216-1GEA
335
SEK
TCIAP2216-1G
TCIAP2216-5G
(1S)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline 98,0 GC, T
(1S)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline
Formel:
C₁₅H₁₅N MW: 209.29 g/mol |
MDL Number:
MFCD08692036 CAS nummer: 118864-75-8 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 87.0 to 91.0 deg-C |
Specific rotation [a]20/D | +10.0 to +14.0 deg (C=0.46, CHCl3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...