2-Phenylethylamine hydroiodide ≥98.0% (by HPLC, total nitrogen)

Leverantör: TCI EUROPE
Varning

Synonyms: Phenethylamine Hydroiodide

P2213-1G P2213-5G
TCIAP2213-1GEA 855 SEK
TCIAP2213-1G TCIAP2213-5G
2-Phenylethylamine hydroiodide ≥98.0% (by HPLC, total nitrogen)
2-Phenylethylamine hydroiodide
Formel: C₈H₁₁N
MW: 249,10 g/mol
CAS nummer: 151059-43-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=095a442bfc44b23c25e5af1a142d0934e944eab5&vid=095a442bfc44b23c25e5af1a142d0934e944eab5&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Water max. 150 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR