1-(4-Propylphenyl)-2,5-di(2-thienyl)-1H-pyrrole ≥98.0%
Leverantör: TCI EUROPE
P2210-200MG
P2210-1G
TCIAP2210-200MGEA
1600
SEK
TCIAP2210-200MG
TCIAP2210-1G
1-(4-Propylphenyl)-2,5-di(2-thienyl)-1H-pyrrole ≥98.0%
1-(4-Propylphenyl)-2,5-di(2-thienyl)-1H-pyrrole
Formel:
C₂₁H₁₉NS₂ |
CAS nummer:
499793-84-9 |
Specifikation testresultat
Purity (GC) | min. 98.0 % |
Melting point | 145.0 to 149.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...