Phenyl trifluoromethyl sulphone ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Phenyl Trifluoromethyl sulphone
P2195-1G
P2195-5G
TCIAP2195-1GEA
550
SEK
TCIAP2195-1G
TCIAP2195-5G
Phenyl trifluoromethyl sulphone ≥98.0%
Phenyl trifluoromethyl sulphone
Formel:
C₇H₅F₃O₂S MW: 210,17 g/mol Kokpunkt: 203°C |
MDL Number:
MFCD00159083 CAS nummer: 426-58-4 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...