Pentafluorophenyl acrylate ≥98.0% stabilised

Leverantör: TCI
Varning

Synonyms: Acrylic Acid Pentafluorophenyl Ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

P2179-1G P2179-5G
TCIAP2179-1GEA 815 SEK
TCIAP2179-1G TCIAP2179-5G
Pentafluorophenyl acrylate ≥98.0% stabilised
Pentafluorophenyl acrylate
Formel: C₉H₃F₅O₂
MW: 238.11 g/mol
Flampunkt: 68°C
Storage Temperature: Kylskåp
MDL Number: MFCD00042330
CAS nummer: 71195-85-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f1baee82d513c9d91f0648d97ae52fe97c3c547d&vid=f1baee82d513c9d91f0648d97ae52fe97c3c547d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR