Piroctone olamine ≥97.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: 1-Hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)-2(1H)-pyridone Ethanolamine, 1-Hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)pyridin-2(1H)-one 2-aminoethanol salt

P2178-1G P2178-5G
TCIAP2178-1GEA 695 SEK
TCIAP2178-1G TCIAP2178-5G
Piroctone olamine ≥97.0% (by HPLC, titration analysis)
Piroctone olamine
Formel: C₁₄H₂₃NO₂
MW: 298,43 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD01690792
CAS nummer: 68890-66-4
EINECS: 272-574-2
Merck Index: 14,07502
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ad31ebac6fc05aad15a3f4696b0aef83ff9184ed&vid=ad31ebac6fc05aad15a3f4696b0aef83ff9184ed&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 134.0 to 138.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR