PD 184,352 ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 2-(2-Chloro-4-iodoanilino)-N-(cyclopropylmethoxy)-3,4-difluorobenzamide, 2-((2-Chloro-4-iodophenyl)amino)-N-(cyclopropylmethoxy)-3,4-difluorobenzamide, PD 184352, CI-1040

P2174-100MG P2174-25MG
TCIAP2174-100MGEA 5950 SEK
TCIAP2174-100MG TCIAP2174-25MG
PD 184,352 ≥98.0%
PD 184,352
Formel: C₁₇H₁₄ClF₂IN₂O₂
MW: 478,7 g/mol
MDL Number: MFCD02683961
CAS nummer: 212631-79-3

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 5.30 to 6.30 %
Melting point 176.0 to 180.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR