4-(Prop-2-yn-1-yl)morpholine ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4-(2-Propyn-1-yl)morpholine, 4-Propargylmorpholine
P2170-200MG
P2170-1G
TCIAP2170-200MGEA
1175
SEK
TCIAP2170-200MG
TCIAP2170-1G
4-(Prop-2-yn-1-yl)morpholine ≥98.0%
4-(Prop-2-yn-1-yl)morpholine
Formel:
C₇H₁₁NO MW: 125,17 g/mol Kokpunkt: 186°C |
MDL Number:
MFCD00510919 CAS nummer: 5799-76-8 UN: 1993 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...