9-Phenylcarbazole-2-boronic acid (contains varying amounts of Anhydride)
Leverantör: TCI
|
Varning
|
P2169-200MG
P2169-1G
TCIAP2169-200MGEA
375
SEK
TCIAP2169-200MG
TCIAP2169-1G
9-Phenylcarbazole-2-boronic acid (contains varying amounts of Anhydride)
9-Phenylcarbazole-2-boronic acid
Formel:
C₁₈H₁₄BNO₂ |
MDL Number:
MFCD22207050 CAS nummer: 1001911-63-2 |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 107.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...