Pidotimod ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
Synonyms:
(R)-3-[[(S)-5-Oxo-2-pyrrolidinyl]carbonyl]-1,3-thiazolidine-4-carboxylic Acid
P2147-5G
P2147-25G
TCIAP2147-5GEA
1600
SEK
TCIAP2147-5G
TCIAP2147-25G
Pidotimod ≥98.0% (by HPLC, titration analysis)
Pidotimod
Formel:
C₉H₁₂N₂O₄S MW: 244,27 g/mol |
MDL Number:
MFCD00867583 CAS nummer: 121808-62-6 Merck Index: 14,07419 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Specific rotation [a]20/D | -131.0 to -141.0 deg (C=2, 5mol/L HCl) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...