Benzoyl(phenyliodonio)(trifluoromethanesulphonyl)methanide ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Shibata Reagent II

P2143-1G P2143-5G
TCIAP2143-1GEA 1225 SEK
TCIAP2143-1G TCIAP2143-5G
Benzoyl(phenyliodonio)(trifluoromethanesulphonyl)methanide ≥98.0%
Benzoyl(phenyliodonio)(trifluoromethanesulphonyl)methanide
Formel: C₁₅H₁₀F₃IO₃S
MW: 454,20 g/mol
Storage Temperature: Frys
CAS nummer: 1443036-49-4
UN: 1759
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d0ac2a698aa4de71d953fc5387fd827475ce4d4&vid=7d0ac2a698aa4de71d953fc5387fd827475ce4d4&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR