Praziquantel ≥98.0% (by HPLC, total nitrogen)
Leverantör: TCI EUROPE
Synonyms:
2-(Cyclohexylcarbonyl)-1,2,3,6,7,11b-hexahydro-4H-pyrazino[2,1-a]isoquinolin-4-one, 2-(Cyclohexanecarbonyl)-2,3,6,7-tetrahydro-1H-pyrazino[2,1-a]isoquinolin-4(11bH)-one
P2125-5G
P2125-25G
TCIAP2125-5GEA
455
SEK
TCIAP2125-5G
TCIAP2125-25G
Praziquantel ≥98.0% (by HPLC, total nitrogen)
Praziquantel
Formel:
C₁₉H₂₄N₂O₂ MW: 312,41 g/mol Storage Temperature: Frys |
MDL Number:
MFCD00058531 CAS nummer: 55268-74-1 EINECS: 259-559-6 Merck Index: 14,07716 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 138.0 to 142.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...