2-Phenoxyacetophenone ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 2-Phenoxy-1-phenylethanone

P2121-1G P2121-5G
TCIAP2121-1GEA 845 SEK
TCIAP2121-1G TCIAP2121-5G
2-Phenoxyacetophenone ≥98.0%
2-Phenoxyacetophenone
Formel: C₁₄H₁₂O₂
MW: 212,25 g/mol
Kokpunkt: 178°C
MDL Number: MFCD00156689
CAS nummer: 721-04-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bffbd538fb776edf1051d4cf6363fddee415c221&vid=bffbd538fb776edf1051d4cf6363fddee415c221&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 71.0 to 75.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR