3-(1,3-Dioxoisoindolin-2-yl)propanoic acid ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 3-Phthalimidopropionic acid, 3-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid

P2101-5G P2101-25G
TCIAP2101-5GEA 630 SEK
TCIAP2101-5G TCIAP2101-25G
3-(1,3-Dioxoisoindolin-2-yl)propanoic acid ≥98.0% (by HPLC, titration analysis)
3-(1,3-Dioxoisoindolin-2-yl)propanoic acid
Formel: C₁₁H₉NO₄
MW: 219,196 g/mol
MDL Number: MFCD00023096
CAS nummer: 3339-73-9
EINECS: 222-083-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db94a54e5ef800cc442afaa637c5dd9ce88aa379&vid=db94a54e5ef800cc442afaa637c5dd9ce88aa379&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 151.0 to 155.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR