2-(2-Pyridyl)benzoxazole ≥97.0% (by GC, titration analysis)

Leverantör: TCI EUROPE

Synonyms: 2-(Pyridin-2-yl)benzo[d]oxazole

P2098-1G P2098-5G
TCIAP2098-1GEA 525 SEK
TCIAP2098-1G TCIAP2098-5G
2-(2-Pyridyl)benzoxazole ≥97.0% (by GC, titration analysis)
2-(2-Pyridyl)benzoxazole
Formel: C₁₂H₈N₂O
MW: 196,21 g/mol
Kokpunkt: 150°C
MDL Number: MFCD00569582
CAS nummer: 32959-62-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38042abb1ef5f8eeece70354c70335d96e4121a5&vid=38042abb1ef5f8eeece70354c70335d96e4121a5&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 103.0 to 109.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR