1-(Phenylsulphonyl)pyrrole ≥98.0%

Leverantör: TCI EUROPE
Varning

P2095-1G P2095-5G
TCIAP2095-1GEA 285 SEK
TCIAP2095-1G TCIAP2095-5G
1-(Phenylsulphonyl)pyrrole ≥98.0%
1-(Phenylsulphonyl)pyrrole
Formel: C₁₀H₉NO₂S
MW: 207,247 g/mol
MDL Number: MFCD00067739
CAS nummer: 16851-82-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd78a8bcd791b8cf748c5ad57fbad4a75a1fce70&vid=dd78a8bcd791b8cf748c5ad57fbad4a75a1fce70&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 87.0 to 91.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR