Paromomycin sulphate ≥94.0%

Leverantör: TCI EUROPE
Varning

Synonyms: (2S,3S,4R,5R,6R)-5-Amino-2-(aminomethyl)-6-(((2R,3S,4R,5S)-5-(((1R,2R,3S,5R,6S)-3,5-diamino-2-(((2S,3R,4R,5S,6R)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-6-hydroxycyclohexyl)oxy)-4-hydroxy-2-(hydroxymethyl)tetrahydrofuran-3-yl)oxy)tetrahydro-2H-pyran-3,4-diol sulphate(1:x)

P2092-1G P2092-5G
TCIAP2092-1GEA 415 SEK
TCIAP2092-1G TCIAP2092-5G
Paromomycin sulphate ≥94.0%
Paromomycin sulphate
Formel: C₂₃H₄₅N₅O₁₄·H₂SO₄
MW: 615,6 g/mol
MDL Number: MFCD00079278
CAS nummer: 1263-89-4
EINECS: 215-031-7
Merck Index: 14,07041
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b959e4a7e3a1b6e2e35c410db4b54acc1a39a7c&vid=1b959e4a7e3a1b6e2e35c410db4b54acc1a39a7c&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 94.0 area%
Specific rotation [a]20/D +47.0 to +55.0 deg (C=1, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR