Phenethyl caffeate ≥98.0%
Leverantör: TCI
|
Varning
|
Synonyms:
3,4-Dihydroxycinnamic Acid Phenethyl Ester, Caffeic Acid Phenethyl Ester, Phenethyl 3-(3,4-dihydroxyphenyl)acrylate, CAPE, Phenethyl 3,4-Dihydroxycinnamate
P2088-25MG
P2088-250MG
TCIAP2088-25MGEA
215
SEK
TCIAP2088-25MG
TCIAP2088-250MG
Phenethyl caffeate ≥98.0%
Phenethyl caffeate
Formel:
C₁₇H₁₆O₄ MW: 284.31 g/mol Storage Temperature: Frys |
MDL Number:
MFCD00866470 CAS nummer: 104594-70-9 |
|
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 127.0 to 131.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...