Pyridine iodine monochloride ≥98.0%

Leverantör: TCI
Varning

Synonyms: Pyridine Iodochloride

P2086-1G P2086-5G
TCIAP2086-1GEA 1150 SEK
TCIAP2086-1G TCIAP2086-5G
Pyridine iodine monochloride ≥98.0%
Pyridine iodine monochloride
Formel: C₅H₅N
MW: 241.46 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD06410987
CAS nummer: 6443-90-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=621ae8eba913ce323c1d1608f7a4df71096c289d&vid=621ae8eba913ce323c1d1608f7a4df71096c289d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (Iodometric Titration) min. 98.0 %
Melting point 133.0 to 137.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR