D-Prolinamide ≥98.0%

Leverantör: TCI EUROPE

Synonyms: H-D-Pro-NH₂, (R)-2-Pyrrolidinecarboxamide, H-D-Pro-NH2

P2083-1G P2083-5G
TCIAP2083-1GEA 590 SEK
TCIAP2083-1G TCIAP2083-5G
D-Prolinamide ≥98.0%
D-Prolinamide
Formel: C₅H₁₀N₂O
MW: 114,15 g/mol
MDL Number: MFCD00153457
CAS nummer: 62937-45-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb46fc89922c8de12a875d1c1f591c04cc0ad3e4&vid=eb46fc89922c8de12a875d1c1f591c04cc0ad3e4&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 99.0 to 103.0 deg-C
Specific rotation [a]20/D +108.0 to +112.0 deg (C=2, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR