Platinum(0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex (19,0-21,5% Pt) (contains 1,3-Divinyltetramethyldisiloxane)
Leverantör: TCI EUROPE
Varning
Synonyms: Karstedt Catalyst, Platinum(0) - 1,3-Divinyltetramethyldisiloxane Complex
P2075-1G P2075-5G
TCIAP2075-1GEA 1725 SEK
TCIAP2075-1G TCIAP2075-5G
Platinum(0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex (19,0-21,5% Pt) (contains 1,3-Divinyltetramethyldisiloxane)
Platinum(0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex
Formel: C₈H₁₈OPtSi₂
MW: 381,485 g/mol
Flampunkt: 28°C
MDL Number: MFCD00151662
CAS nummer: 68478-92-2
EINECS: 270-844-4
UN: 1993
ADR: 3,III
Order Now
Specifikation testresultat
Appearance Light yellow to Yellow to Green clear liquid
Content (Platinum) 19.00 to 21.50 %