Pramipexole dihydrochloride monohydrate ≥98.0% (by HPLC, total nitrogen)

Leverantör: TCI EUROPE
Varning

Synonyms: (S)-2-Amino-4,5,6,7-tetrahydro-6-(propylamino)benzothiazole Dihydrochloride, (S)-N6-Propyl-4,5,6,7-tetrahydrobenzo[d]thiazole-2,6-diamine dihydrochloride hydrate

P2073-100MG P2073-1G
TCIAP2073-100MGEA 350 SEK
TCIAP2073-100MG TCIAP2073-1G
Pramipexole dihydrochloride monohydrate ≥98.0% (by HPLC, total nitrogen)
Pramipexole dihydrochloride monohydrate
Formel: C₁₀H₂₁Cl₂N₃OS
MW: 302,26 g/mol
MDL Number: MFCD02183927
CAS nummer: 191217-81-9
Merck Index: 14,07705
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f3c892e4615a2a670bb6ca1aa7fcb5b4506b2f5&vid=2f3c892e4615a2a670bb6ca1aa7fcb5b4506b2f5&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Specific rotation [a]20/D -65.0 to -71.0 deg (C=1,MeOH)
Water 5.0 to 7.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR