1-Phenyl-1,2,3,4-tetrahydroisoquinoline 98,0 GC, T
Leverantör: TCI
Synonyms:
1,2,3,4-Tetrahydro-1-phenylisoquinoline
P2056-1G
P2056-5G
TCIAP2056-1GEA
470
SEK
TCIAP2056-1G
TCIAP2056-5G
1-Phenyl-1,2,3,4-tetrahydroisoquinoline 98,0 GC, T
1-Phenyl-1,2,3,4-tetrahydroisoquinoline
Formel:
C₁₅H₁₅N MW: 209.29 g/mol |
MDL Number:
MFCD02179241 CAS nummer: 22990-19-8 |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 98.0 to 102.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...