Palonosetron hydrochloride ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI

Synonyms: (S)-2-((S)-Quinuclidin-3-yl)-2,3,3a,4,5,6-hexahydro-1H-benzo[de]isoquinolin-1-one hydrochloride

P2051-200MG P2051-1G
TCIAP2051-200MGEA 1025 SEK
TCIAP2051-200MG TCIAP2051-1G
Palonosetron hydrochloride ≥98.0% (by HPLC, titration analysis)
Palonosetron hydrochloride
Formel: C₁₉H₂₄N₂O
MW: 332.87 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD07370072
CAS nummer: 135729-62-3
Merck Index: 14,06997
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de35a18eb9057a99771b19d27ef81e2718e6e967&vid=de35a18eb9057a99771b19d27ef81e2718e6e967&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -88.0 to -93.0 deg (C=1, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR