5-Phenyl-3-isoxazolecarboxylic acid ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 5-Phenylisoxazole-3-carboxylic acid

P2020-1G P2020-5G
TCIAP2020-1GEA 750 SEK
TCIAP2020-1G TCIAP2020-5G
5-Phenyl-3-isoxazolecarboxylic acid ≥98.0% (by HPLC, titration analysis)
5-Phenyl-3-isoxazolecarboxylic acid
Formel: C₁₀H₇NO₃
MW: 189,1700 g/mol
MDL Number: MFCD00464221
CAS nummer: 14441-90-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76930d75a5a43a111a28540604eca0c495d181e6&vid=76930d75a5a43a111a28540604eca0c495d181e6&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 158.0 to 162.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR