4-(4-Piperidyl)pyridine ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 4-(4-Pyridyl)piperidine, 4-(Piperidin-4-yl)pyridine

P2012-1G P2012-5G
TCIAP2012-1GEA 2425 SEK
TCIAP2012-1G TCIAP2012-5G
4-(4-Piperidyl)pyridine ≥98.0%
4-(4-Piperidyl)pyridine
Formel: C₁₀H₁₄N₂
MW: 162,24 g/mol
Kokpunkt: 292°C
MDL Number: MFCD05863877
CAS nummer: 581-45-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afdbd33f6d41c045b4ad9d06b41c2799eba6b6f1&vid=afdbd33f6d41c045b4ad9d06b41c2799eba6b6f1&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 84.0 to 88.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR